Learn More
Selleck Chemical LLC Sodium nitroprusside S4493-1g
SDP
Supplier:  Selleck Chemical LLC S44931G
Encompass_Preferred
Sodium nitroprusside (SNP Ro 21-2498) is a generically available and rapid-acting intravenous (IV) vasodilator that dilates cerebral vessels and increases internal carotid (ICA) flow
Catalog No. 50-311-0208