Learn More
Selleck Chemical LLC Sodium Nitroprusside Dihydrate S4059-50mg
SDP
Supplier:  Selleck Chemical LLC S405950MG
Encompass_Preferred
Sodium Nitroprusside Dihydrate is a potent vasodilator working through releasing NO spontaneously in blood
Catalog No. 50-310-9576