Learn More
Apexbio Technology LLC Sodium Orthovanadate 13721-39-6 5g
SDP
Supplier:  Apexbio Technology LLC A85245000
Encompass_Preferred
Sodium orthovanadate (CAS 13721-39-6) is an inorganic compound widely utilized as a broad-spectrum inhibitor of protein tyrosine phosphatases (PTPs) alkaline phosphatase (ALP) and ATPase It acts as a reversible competitive inhibitor its effects can be reversed by chelating agents such as EDTA or by dilution Sodium orthovanadate is routinely employed to maintain tyrosine phosphorylation states in cells cell lysates and in protein tyrosine kinase assays Additionally it enables the selective inhibition of PTPs distinct from protein phosphoseryl and phosphothreonyl phosphatases which are targeted by agents such as fluoride and EDTA
Catalog No. 50-000-95202