Learn More
Selleck Chemical LLC Sodium orthovanadate S2000-1g
SDP
Supplier:  Selleck Chemical LLC S20001G
Encompass_Preferred
Sodium orthovanadate is an alkaline phosphatase and (Na K)-ATPase inhibitor with IC50 of 10 M
Catalog No. 50-311-0059