Learn More
Enzo Life Sciences Sodium orthovanadate (5g). CAS: 13721-39-6
SDP
Supplier:  Enzo Life Sciences ALX400032G005
Encompass_Preferred
Potent inhibitor of alkaline phosphatase and tyrosine phosphates. Regarding decavanadate: across the pH range of 1-13, simple aqueous solutions of [VO4]3- ion involve a dozen or more ionic species, both monomeric and oligomeric, whose abundances depend strongly on pH and [VO4]3- concentration. At pH 3.75 a stable complex can be prepared which contains ten vanadium atoms and hence is a decavanadate ion. When diluted to pH 7.2, the decavanadate species has a half-life time of about 2 hours. Fohr et al., also provide directions for generating control solutions of mixed vanadate complexes in which decavanadate is absent and monomeric orthovanadate is present. Decavanadate inhibits inositol 1,4,5-trisphosphate- (IP3) induced calcium release from permeabilized endocrine cells and the binding of [3H]IP3 to its receptor in cerebellar and adrenal cortical membranes. Alternative name: Sodium vanadate. Purity: ≥90% (dry basis titration) . Solubility: Soluble in water (100mg/ml).
Catalog No. 50-200-9074