Learn More
Selleck Chemical LLC Sodium orthovanadate 100mg 13721-39-6
SDP
Supplier:  Selleck Chemical LLC S2000100MG
Encompass_Preferred
Sodium orthovanadate is an alkaline phosphatase and (Na,K)-ATPase inhibitor with IC50 of 10 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-573-0