Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium Orthovanadate Activ
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 5086050004
Encompass
A depolymerized vanadate prepared from Sodium Orthovanadate (Cat. No. 567540) that exhibits significant enhanced ability to inhibit endogenous phosphatases in a competitive manner. Preserves the phosphorylation state in cells and cell lysates. Completely reversible with EDTA and dilution. Recommended concentration in lysis buffer ~ 1 mM. Available as 5 ml or as a set of 3 x 1 ml.
Catalog No. 50-290-1305