Learn More
New England Biolabs, Inc. Sodium Orthovanadate (Vanadate) – 5 ml
SDP
Supplier:  New England Biolabs, Inc. P0758L
Encompass
Sodium Orthovanadate (Vanadate) is a general competitive inhibitor for protein phosphotyrosyl phosphatases. The inhibition by Sodium Orthovanadate is reversible upon the addition of EDTA or by dilution.
Catalog No. 50-995-099