Learn More
Cayman Chemical SodIum Orthovnadate 50g
SDP
Supplier:  Cayman Chemical 3432850 G
Encompass_Preferred
A protein tyrosine phosphatase inhibitor; inhibits autocrine growth of A549 lung, A498 kidney, and DU145 prostate cancer cells from 1-20 µM; protects mice from radiation-induced hematopoietic and gastrointestinal syndromes at 20 mg/kg; reduces postprandial glucose levels and increases glucose tolerance in a rat model of streptozotocin-induced diabetes; decreases total and cortical infarct size in a rat model of transient MCAO-induced ischemia-reperfusion injury
Catalog No. 50-295-0381