Learn More
Cayman Chemical SodIum Oxamate 25g
SDP
Supplier:  Cayman Chemical 1905725 G
Encompass_Preferred
A derivative of pyruvate that inhibits the conversion of pyruvate to lactate via lactate dehydrogenase, thus disrupting glycolysis; studied as an inhibitor of glycolysis in various tumors
Catalog No. 50-298-3487