Learn More
Strem, An Ascensus Company 10g. Sodium oxide/sodium on alumina, Olefin Isomerization Catalyst (Na2O 11.5-13.5%, Na 1.8-3.0%).
SDP
Supplier:  Strem, An Ascensus Company 11100710G
Encompass
10g. Sodium oxide/sodium on alumina, Olefin Isomerization Catalyst (Na2O 11.5-13.5%, Na 1.8-3.0%). . Molecular Weight: . Molecular Formula: Na2O/Na. Color/form: white solid Strem# 11-1007. http://www.strem.com/catalog/v/11-1007/
Catalog No. 50-366-8