Learn More
Research Products International Corp Sodium Oxalate, 100 Grams

Supplier: Research Products International Corp S24500100.0
- CAS Number: 67-76-0
- Molecular Formula: C2Na2O4
- Molecular Weight: 134
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.