Learn More
Selleck Chemical LLC Sodium palmitate E2900-250MG
SDP
Supplier:  Selleck Chemical LLC E2900250MG
Encompass_Preferred
Sodium palmitate(Palmitic acid sodium) is a common saturated fatty acid and produced during fatty acid synthesis Sodium palmitate enhances lipogenesis and cellular steatosis in various cell lines
Catalog No. 50-000-40542