Learn More
Alkali Scientific Sodium Perchlorate, ACS Grade, 10 Kg
SDP
Supplier:  Alkali Scientific UMP005CP
Encompass_Preferred
Sodium Perchlorate, ACS Grade, is a high-purity inorganic salt available in a 10 kg quantity. It is commonly used as an oxidizing agent in a wide range of laboratory and industrial applications. This chemical is ideal for high-temperature reactions and the synthesis of perchloric acid, which is used in various chemical processes. Sodium perchlorate’s stable oxidizing properties make it essential for numerous applications, including chemical analysis, organic synthesis, and in reactions requiring controlled oxidation. It is widely utilized in laboratories for both research and educational purposes, ensuring consistent and reliable results.
Catalog No. 50-006-61734