Learn More
Alkali Scientific Sodium Perchlorate, ACS Grade, 500 G
SDP
Supplier:  Alkali Scientific UMP002CP
Encompass_Preferred
Sodium Perchlorate, ACS Grade, is a highly purified inorganic salt commonly used as an oxidizing agent in chemical reactions. This 500g package is designed for laboratory applications requiring precise and consistent oxidation reactions, such as the synthesis of perchlorate salts and the degradation of organic compounds. Sodium perchlorate is used in various chemical processes, including analytical chemistry and environmental testing, where oxidation is required to break down contaminants. It is an essential reagent in inorganic chemistry and research labs where high-purity chemicals are critical to obtaining reliable results in oxidation and chemical synthesis processes.
Catalog No. 50-006-60573