Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium perchlorate ACS rea25KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 4102412.5KG
Encompass
Sodium perchlorate is a water soluble perchlorate salt. It is prepared by electrolyzing aqueous sodium chlorate SOLUTIONon. It is a strong oxidizing agent. On impregnating porous silicon with sodium perchlorate it forms an explosive composite which has been investigated.
Catalog No. 50-289-8319