Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium perchlorate monohyd1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 718531KG
Encompass
Sodium perchlorate monohydrate (NaClO4A H2O) is a hydrated sodium salt. Its crystal structure has been investigated by three-dimensional single crystal x-ray diffraction studies. Its crystals belong to the monoclinic crystal system having space group C2/c.
Catalog No. 50-289-9174