Learn More
Hach Company Sodium Periodate Powder Pillows for Manganese, 10 mL, pk/100
Supplier:  Hach Company 2107769
Encompass
For determination of manganese by the periodate oxidation method. EPA approved for reporting wastewater analyses (digestion required). Hach method 8034. Sample size: 10 mL Pack of 100 Powder Pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-5423