Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium phenylbutyrate, 1716-12-7, MFCD00800247, 100 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SML0309100MG
Encompass
Empirical Formula (Hill Notation): C10H11NaO2, Molecular Weight: 186.18, ≥98% (HPLC), storage temp: room temp, Synonym: 4-PBA, 4-Phenylbutyric acid, 4-phenylbutyrate, Buphenyl, Sodium 4-phenylbutyrate, TriButyrate.
Catalog No. NC0784773