Learn More
Apexbio Technology LLC Sodium phosphate dibasic 7558-79-4 500g
SDP
Supplier:  Apexbio Technology LLC B7293500000
Encompass_Preferred
Sodium phosphate dibasic (CAS 7558-79-4) is an inorganic compound commonly used as a buffering reagent It acts by participating in buffering systems accepting or releasing hydrogen ions to maintain stable pH levels in aqueous solutions Sodium phosphate dibasic is widely utilized in maintaining physiological pH conditions during biochemical reaction setups molecular biology procedures and cell-culture media preparations It is frequently employed in enzymatic assays nucleic acid research protein extraction and other biochemical applications requiring stringent control over reaction environment stability
Catalog No. 50-000-90996