Learn More
Alkali Scientific Sodium Phosphate Dibasic, Anhydrous, ACS Grade, 1 Kg
SDP
Supplier:  Alkali Scientific EDI006CP
Encompass_Preferred
Sodium Phosphate Dibasic, Anhydrous, ACS Grade, 1 Kg is a high-purity reagent used for a variety of laboratory applications, including pH adjustment and buffering. It is an essential chemical for analytical and biological processes, offering a stable buffering capacity at a neutral pH. The anhydrous form provides flexibility for precise concentration control in formulations and reactions. This reagent is commonly used in the preparation of phosphate buffers, where it helps maintain the pH for sensitive processes like chromatography and molecular biology assays. Its reliable performance makes it an ideal choice for laboratories conducting precise chemical and biological analysis.
Catalog No. 50-006-60187