Learn More
Alkali Scientific Sodium Phosphate Dibasic, Anhydrous, ACS Grade, 25000 G
SDP
Supplier:  Alkali Scientific EDI007CP
Encompass_Preferred
Sodium Phosphate Dibasic, Anhydrous, ACS Grade, 25,000 G is a high-grade reagent in bulk packaging, ideal for laboratories requiring large quantities of phosphate buffer or solutions. This chemical is essential in analytical chemistry for preparing phosphate buffers that maintain stable pH levels in various processes such as titration, chromatography, and molecular biology. The anhydrous form ensures that the reagent remains stable and easy to dissolve in water, offering precise control over solution concentrations. It is commonly used in large-scale applications including pharmaceutical production, environmental analysis, and chemical manufacturing, providing a cost-effective and reliable buffer system.
Catalog No. 50-006-60383