Learn More
Alkali Scientific Sodium Phosphate Dibasic Heptahydrate, 1 Kg
SDP
Supplier:  Alkali Scientific DIB001CP
Encompass_Preferred
Sodium Phosphate Dibasic Heptahydrate, 1 Kg, ACS Grade, is a laboratory-grade reagent that is commonly used in the preparation of phosphate buffer solutions. The heptahydrate form of sodium phosphate provides a stable, water-soluble compound that is ideal for adjusting pH in various chemical and biological applications. This reagent is often used in microbiology, biochemistry, and environmental testing for its ability to maintain pH stability, ensuring optimal conditions for reactions. It is particularly important in applications like chromatography, sample preparation, and routine lab tests where precise pH control is critical for accurate results.
Catalog No. 50-006-61846