Learn More
Alkali Scientific Sodium Phosphate Dibasic Heptahydrate, 10 Kg
SDP
Supplier:  Alkali Scientific DIB002CP
Encompass_Preferred
Sodium Phosphate Dibasic Heptahydrate, 10 Kg, ACS Grade, is a bulk reagent designed for large-scale laboratory and industrial use. This compound is essential in preparing phosphate buffer solutions that require precise pH control in chemical, biological, and pharmaceutical applications. The heptahydrate form provides a stable, easy-to-dissolve compound that ensures accurate buffering capacity. This larger 10 Kg packaging is ideal for high-volume labs and manufacturing processes that routinely require large quantities of buffer solutions, making it an economical and reliable choice for laboratories engaged in extensive research or production work.
Catalog No. 50-006-61123