Learn More
Alkali Scientific Sodium Phosphate Dibasic Heptahydrate, 25 Kilograms
SDP
Supplier:  Alkali Scientific DIB006CP
Encompass_Preferred
Sodium Phosphate Dibasic Heptahydrate is a high-purity reagent used primarily in laboratory applications for preparing phosphate buffers and maintaining stable pH conditions during various experiments. The 25 kg package is ideal for large-scale production of buffer solutions or for industrial applications requiring consistent and reliable buffering capacity. Sodium phosphate dibasic is often used in biochemical assays, protein purification, and nucleic acid analysis. Its stable buffering capabilities make it an essential component for experiments involving enzyme reactions, chromatography, and various biological assays. The high purity of this reagent ensures that experiments yield accurate and reproducible results.
Catalog No. 50-006-60208