Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium phosphate dibasic p5KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 716405KG
Encompass
Sodium phosphate is widely used buffer in molecular biology biochemistry and chromatographic studies. It exists in three forms monobasic (NaH2PO4) dibasic (Na2HPO4) and tribasic (Na3PO4). Monobasic and dibasic forms in varying ratios are commonly used to compose the neutral sodium phosphate buffer SOLUTIONon.
Catalog No. 50-289-9855