Learn More
Alkali Scientific Sodium Phosphate, Monobasic, Anhydrous, 1 Kg
SDP
Supplier:  Alkali Scientific SOA001CP
Encompass_Preferred
Sodium Phosphate, Monobasic, Anhydrous, 1 Kg, ACS Grade, is a high-purity reagent used in laboratories to prepare phosphate buffer solutions. The anhydrous form of sodium phosphate ensures that it remains stable and easy to dissolve, offering precise control over buffer concentrations. This reagent is widely used in analytical chemistry, biochemistry, and molecular biology for processes that require a stable pH environment, such as chromatography, electrophoresis, and enzyme assays. It is commonly employed in biological and chemical research to prepare solutions that support cellular activities or stabilize proteins during analysis.
Catalog No. 50-006-61031