Learn More
Alkali Scientific Sodium Phosphate, Monobasic, Anhydrous, 5 Kg
SDP
Supplier:  Alkali Scientific SOA004CP
Encompass_Preferred
Sodium Phosphate Monobasic Anhydrous (5 Kg) is a high-purity reagent widely used in laboratory applications for buffer preparation, pH control, and analytical chemistry. This anhydrous form of sodium phosphate ensures minimal moisture interference and provides a more stable, concentrated form of the reagent for precise pH adjustments. The 5 Kg packaging is ideal for labs with medium to high throughput, where consistent preparation of phosphate buffers is required. It is commonly used in various biochemical and molecular biology processes, including enzyme assays, chromatography, and DNA extraction, where maintaining pH stability is critical for accurate and reproducible results.
Catalog No. 50-006-61146