Learn More
Alkali Scientific Sodium Phosphate Monobasic, Monohydrate, ACS Grade, 5 Kg
SDP
Supplier:  Alkali Scientific EMO008CP
Encompass_Preferred
Sodium Phosphate Monobasic, Monohydrate (ACS Grade) is a premium reagent used to prepare acidic phosphate buffers in laboratory and industrial applications. The 5 kg quantity is well-suited for high-volume production of buffer solutions for routine testing and experimental procedures. Sodium phosphate monobasic is widely used for pH adjustment and buffering in analytical chemistry, biochemistry, and molecular biology. It is essential in preparing buffers for enzyme assays, chromatography, and nucleic acid studies, ensuring accurate pH control and stability throughout experiments. This ACS-grade reagent provides the necessary precision and reliability required in high-quality analytical and research applications.
Catalog No. 50-006-59916