Learn More
Alkali Scientific Sodium Phosphate Monobasic Monohydrate [Sodium dihydrogen phosphate monohydrate], 3 Kg
SDP
Supplier:  Alkali Scientific EMO002CP
Encompass
Sodium Phosphate Monobasic Monohydrate, 3 Kg is an essential laboratory reagent used to prepare phosphate buffer solutions and control pH in biochemical, pharmaceutical, and analytical applications. The monohydrate form ensures the appropriate hydration of the compound, which is important for maintaining the stability and accuracy of the buffer solutions. It is commonly used in laboratory experiments that require a specific acidic pH for enzymatic reactions, protein analysis, and DNA/RNA work. The 3 Kg size is ideal for laboratories that perform medium to large-scale experiments or need to replenish buffer solutions regularly. It is a versatile and indispensable reagent in many laboratory protocols.
Catalog No. 50-006-59831