Learn More
Alkali Scientific Sodium Phosphate Monobasic Monohydrate [Sodium dihydrogen phosphate monohydrate], 5 Kg
SDP
Supplier:  Alkali Scientific EMO004CP
Encompass_Preferred
Sodium Phosphate Monobasic Monohydrate, 5 Kg is a high-concentration reagent used for the preparation of phosphate buffer solutions in laboratory applications. This product is commonly used in biochemistry, molecular biology, and pharmaceutical testing for pH adjustments in a variety of analytical procedures. The monohydrate form provides the correct level of hydration needed to ensure buffer stability and accuracy. The 5 Kg size is well-suited for larger-scale laboratories or production settings that require high quantities of reagent for routine use. Sodium phosphate monobasic is essential for maintaining pH during enzyme assays, protein purification, and other experiments where consistent acidity is required.
Catalog No. 50-006-59809