Learn More
Apexbio Technology LLC Sodium phosphate monobasic 7558-80-7 250g
SDP
Supplier:  Apexbio Technology LLC B7779250000
Encompass_Preferred
Sodium phosphate monobasic (7558-80-7) is an inorganic sodium salt frequently used as an ionic buffering agent It is designed to modulate pH and osmotic balance in aqueous systems thereby supporting stable conditions in biological and chemical experiments Sodium phosphate monobasic exerts its functional role primarily through pH regulation No direct molecular target or activity metric is reported In biological research sodium phosphate monobasic is widely utilized to prepare buffer solutions for cell culture enzyme assays and molecular biology applications where precise pH control is needed It is also employed in studies examining gastrointestinal fluid dynamics and electrolyte balance Based on these properties sodium phosphate monobasic holds research potential in physiological studies buffer preparation and water treatment applications
Catalog No. 50-224-0654