Learn More
Medchemexpress LLC Sodium Potassium ATPase Antibody | 113 kDa | 50 UL

Supplier: Medchemexpress LLC HYP8053750UL
The Sodium Potassium ATPase Antibody is a non-conjugated, rabbit polyclonal IgG antibody, targeting alpha 1 Sodium Potassium ATPase. Affinity purified, it serves as a reliable loading control antibody for various research applications.
- Targets alpha 1 Sodium Potassium ATPase
- Host: Rabbit
- Clonality: Polyclonal
- Species reactivity: Human, Mouse, Rat
- Predicted molecular weight: 113 kDa
- Applications include Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC)
- Affinity purified
- Non-conjugated
- Isotype: IgG
- Store at -20°C for 1 year
- Avoid repeated freeze/thaw cycles
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.