Learn More
Medchemexpress LLC Sodium Potassium ATPase Antibody | Predicted band size: 113 kDa; Observed band size: 100 kDa | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP8053710UL
Encompass_Preferred
Sodium Potassium ATPase Antibody is a non-conjugated, Rabbit-originated IgG polyclonal antibody that targets alpha 1 Sodium Potassium ATPase. It can be used as a loading control antibody. This antibody is crucial for research on the sodium-potassium pump, an integral membrane protein responsible for maintaining ion gradients across cell membranes, essential for osmoregulation, transport of molecules, and electrical excitability.
  • Targets alpha 1 Sodium Potassium ATPase.
  • Rabbit-originated IgG polyclonal antibody.
  • Can be used as a loading control antibody.
  • Validated for applications such as Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC).
  • Reactivity observed in human, mouse, and rat species.
  • Supplied in a stable formulation designed for 1 year storage at -20°C.
  • For research use only.
Catalog No. 50-003-96220