Learn More
Selleck Chemical LLC Sodium propionate E4022-1G
SDP
Supplier:  Selleck Chemical LLC E40221G
Encompass_Preferred
Sodium propionate (sodium propanoate) a short chain fatty acid is a white crystalline salt derived from propionic acid with anti-inflammatory properties Sodium propionate is used as a supplement in transgalactic-oligosaccharides (TOS) medium for the isolation of bifidobacteria and as a supplement in the SCFA (short-chain fatty acid) diet prepared for mice
Catalog No. 50-000-39004