Learn More
Selleck Chemical LLC Sodium pyrophosphate 10H20 E4001-1G
SDP
Supplier:  Selleck Chemical LLC E40011G
Encompass_Preferred
Sodium pyrophosphate decahydrate acts as a buffering agent a chelator and a hydrated salt nucleating agent Sodium pyrophosphate decahydrate can also be used to a raw material for synthetic laundry scent additives
Catalog No. 50-000-43045