Learn More
Selleck Chemical LLC Sodium pyrophosphate 10H21 E4001-250MG
SDP
Supplier:  Selleck Chemical LLC E4001250MG
Encompass_Preferred
Sodium pyrophosphate decahydrate acts as a buffering agent a chelator and a hydrated salt nucleating agent Sodium pyrophosphate decahydrate can also be used to a raw material for synthetic laundry scent additives
Catalog No. 50-000-42530