Learn More
ILEX LIFE SCIENCES LLC PAN Biotech Sodium Pyruvate 100 mM Solution cell culture media supplement 600 ML 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0443100X6
Encompass_Preferred
PAN-Biotech Sodium Pyruvate Solution 100 mM cell culture media supplement 6 x 100 ml DESCRIPTION Sodium Pyruvate Solution finds application in cell culture as an easily accessible carbohydrate source for cells Sodium pyruvate is also involved with amino acid metabolism and initiates the Kreb s cycle In general the 100 mM solution should be diluted 1 100 for most cell culture applications COMPOSITION 11 050 00 mg/L Sodium Pyruvate CAS NO 113-24-6 LIQUID/POWDER Liquid STERILE Yes STORAGE TEMPERATURE 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80414