Learn More
Selleck Chemical LLC Sodium Pyruvate E2902-25MG
SDP
Supplier:  Selleck Chemical LLC E290225MG
Encompass_Preferred
Sodium Pyruvate(Sodium 2-oxopropanoate) is a compound produced in the glycolytic pathway Sodium Pyruvate is a free radical scavenger that can scavenge ROS
Catalog No. 50-000-40541