Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium (S)-2-hydroxypropanoate-d3 | 285979-84-2 | 98.3% | 115.08 | C3H2D3NaO3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYW040233S10MG

Encompass_Preferred

L-Lactic acid-d3 sodium is the deuterium-labeled sodium salt of L-lactic acid ((S)-2-hydroxypropanoic acid). It is supplied as a high-purity, isotopically labeled building block and tracer reagent for analytical, metabolic, and research applications where deuterium labeling aids tracking and quantitation.

  • Deuterium-labeled (d3) sodium salt for isotopic tracing.
  • Purity 98.31%.
  • Molecular formula C3H2D3NaO3; molecular weight 115.08.
  • Available in small pack sizes for research use (e.g., 10 mg).
  • Store sealed at 4°C; in solution, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-08739


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.