Learn More
Medchemexpress LLC Sodium (S)-3-amino-3-carboxypropanoate hydrate | 323194-76-9 | MFCD00152960 | >98.0% | 173.10 g/mol | C4H8NNaO5 | 1 KG

Supplier: Medchemexpress LLC HYW1409001KG
Sodium (S)-3-amino-3-carboxypropanoate hydrate (Sodium L-aspartate monohydrate) is a white to off-white solid biochemical reagent intended for research use. It is used in biochemical assays and as a component of cardioplegia solutions in bypass surgery protocols. Product is supplied with quality documentation and recommended storage conditions for laboratory use.
- High purity (>98.0%).
- Molecular weight 173.10 g/mol.
- White to off-white solid appearance.
- Available in multiple pack sizes, including 1 KG.
- Supplied with certificate of analysis and safety data sheet.
- Recommended storage: 4°C, sealed and away from moisture.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.