Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium salicylate, 54-21-7, MFCD00002440, 500 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S3007500G
Encompass
Linear Formula: HOC6H4COONa, Molecular Weight: 160.10, ReagentPlus, ≥99.5%, Synonym: 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt, Sodium 2-hydroxybenzoate. Sodium salicylate has been used: to induce tinnitus, hyperacusis-like behavior in rats and infused directly into amygdala to measure local field potential.
Catalog No. NC1082260