Learn More
Strem, An Ascensus Company CAS# 822-16-2. 1kg. Sodium stearate, tech. gr.. MFCD00036404
SDP
Supplier:  Strem, An Ascensus Company 9311801000G
Encompass
CAS# 822-16-2. 1kg. Sodium stearate, tech. gr.. MFCD00036404. Molecular Weight: 306.49. Molecular Formula: Na(OOCC17H35). Color/form: white pwdr. Strem# 93-1180. http://www.strem.com/catalog/v/93-1180/
Catalog No. 50-901-13901