Learn More
Medchemexpress LLC Sodium stearyl fumar 50g
SDP
Supplier:  Medchemexpress LLC HYW09955850G
Encompass_Preferred
Sodium stearyl fumarate is a tablet lubricant Sodium stearyl fumarate improves tablet compressibility[1]
Catalog No. 50-002-05870