Learn More
Cayman Chemical SodIum StIbogluconate 1g
SDP
Supplier:  Cayman Chemical 160381 G
Encompass_Preferred
A pentavalent antimony derivative that is highly active against Leishmania amastigotes in macrophages
Catalog No. 50-296-8844