Learn More
Alkali Scientific Sodium Sulfate, Anhydrous, ACS Grade, 5 Kg
SDP
Supplier:  Alkali Scientific UMS003CP
Encompass_Preferred
Sodium Sulfate, Anhydrous, ACS Grade is a high-quality chemical reagent available in a 5kg quantity, suitable for laboratory and industrial applications. This anhydrous form of sodium sulfate is used as a drying agent and in the preparation of buffer solutions. It is commonly used in analytical chemistry, particularly in gravimetric analysis, and in applications requiring precise ionic strength. Sodium Sulfate has a variety of uses in the preparation of chemical compounds, as well as in manufacturing detergents, glass, and other industrial materials. This ACS Grade reagent meets the stringent standards for accuracy and purity in chemical analysis and research environments.
Catalog No. 50-006-61797