Learn More
Alkali Scientific Sodium Sulfate, Anhydrous, ACS Grade, 500 G
SDP
Supplier:  Alkali Scientific UMS002CP
Encompass_Preferred
Sodium Sulfate, Anhydrous, ACS Grade, is available in a 500g bottle, providing a pure and stable source of sodium sulfate for laboratory use. Sodium sulfate is commonly used in laboratory applications such as reagent preparation, chemical synthesis, and as a drying agent for organic solvents. The anhydrous form ensures no moisture content, making it suitable for sensitive experiments and procedures requiring precise conditions. The ACS grade ensures that the product meets strict purity standards for accurate and reliable scientific research, making it an essential chemical reagent in both academic and industrial laboratories.
Catalog No. 50-006-61731