Learn More
Alkali Scientific Sodium Sulfate, Anhydrous, ACS Grade, 1 Kg
Supplier: Alkali Scientific UMS001CP
Sodium Sulfate, Anhydrous, ACS Grade, is a white, crystalline powder that is used in laboratories for a variety of applications. This 1 kg bottle provides sodium sulfate in its anhydrous form, ensuring that there is no moisture content, which makes it ideal for precise chemical reactions, analytical procedures, and laboratory testing. Sodium sulfate is often used in the preparation of reagents, in the synthesis of other chemicals, and in the drying of organic compounds. The ACS grade guarantees that the product meets the highest standards of purity, making it suitable for use in research, chemical analysis, and industrial applications.