Learn More
Medchemexpress LLC Sodium sulfate decahydrate | 7727-73-3 | MFCD00149189 | 322.19 g·mol⁻¹ | Na2SO4·10H2O | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01987750G
Encompass_Preferred
Sodium sulfate decahydrate (Na2SO4·10H2O, CAS 7727-73-3) is a commonly used inorganic reagent for chemical and biochemical research. It is used as a salting-out agent, to assist sulfation of phenolic compounds, and in studies of ionic strength, crystallization, and formulation.
  • Useful reagent for salting-out and crystallization studies.
  • Promotes sulfation of phenolic compounds in biochemical assays.
  • Formula Na2SO4·10H2O with molecular weight ≈322.19 g·mol⁻¹.
  • White to off-white crystalline solid, soluble in water (≈200 mg/mL).
  • Available in small pack sizes suitable for bench use.
  • Store sealed and protected from moisture; keep at 4 °C for neat material.
Catalog No. 50-003-00288