Learn More
Strem, An Ascensus Company CAS# 865-48-5. 100G. Sodium t-butoxide, min. 98%. MFCD00040575
SDP
Supplier:  Strem, An Ascensus Company 931022100G
Encompass
CAS# 865-48-5. 100G. Sodium t-butoxide, min. 98%. MFCD00040575. Molecular Formula: C4H9ONa. Molecular Weight: 96.11. Color/Form: white to off-white pwdr. Stability: moisture sensitive. Strem# 93-1022. www.strem.com/catalog/v3/93-1022/
Catalog No. 50-196-3544